accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 870703-75-6

NNHBrF

CAS RN 870703-75-6

IUPAC Name: 1-[(4-bromo-2-fluorophenyl)methyl]piperazine
Synonyms: 1-[(4-bromo-2-fluorophenyl)methyl]piperazine
Molecular Weight (g/mol): 273.15
Molecular Formula: C11H14BrFN2
InChi Key: NRCBEINBLSTDHM-UHFFFAOYSA-N
SMILES: FC1=C(CN2CCNCC2)C=CC(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results