accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFNH2HCl

CAS RN 870562-90-6

IUPAC Name: 1-(4-fluoro-2-methoxyphenyl)methanamine hydrochloride
Synonyms: 1-(4-fluoro-2-methoxyphenyl)methanamine hydrochloride
Molecular Weight (g/mol): 191.63
Molecular Formula: C8H11ClFNO
InChi Key: GLKUYMNEPUILTR-UHFFFAOYSA-N
SMILES: Cl.COC1=C(CN)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results