accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 850568-45-5

BClNH2HOOHHCl

CAS RN 850568-45-5

IUPAC Name: (3-amino-4-chlorophenyl)boronic acid hydrochloride
Synonyms: 3-amino-4-chlorophenylboronic acid hydrochloride
Molecular Weight (g/mol): 207.85
Molecular Formula: C6H8BCl2NO2
InChi Key: MTMLYYSBEQYCDJ-UHFFFAOYSA-N
SMILES: Cl.NC1=C(Cl)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results