accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOOHNSOONHSN

CAS RN 85-73-4

IUPAC Name: 2-({4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}carbamoyl)benzoic acid
Synonyms: phthalylsulfathiazole phthalazole sulfaphthalazole thalistatyl thalazole phthalidin phthalylnorsulfazole ftalil-esteve ftalazol sulphaphthalyl
Molecular Weight (g/mol): 403.43
Molecular Formula: C17H13N3O5S2
InChi Key: PBMSWVPMRUJMPE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results