accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 847818-74-0

BOOH3CCH3H3CH3CNNH3C

CAS RN 847818-74-0

IUPAC Name: 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 208.07
Molecular Formula: C10H17BN2O2
InChi Key: HLXOVAMYQUFLPE-UHFFFAOYSA-N
SMILES: CN1N=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results