accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNNSClOHOOHOOHOOHO(Z)(Z)

CAS RN 84-02-6

IUPAC Name: bis((2Z)-but-2-enedioic acid); 2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]-10H-phenothiazine
Synonyms: bis(maleic acid); prochlorperazine
Molecular Weight (g/mol): 606.09
Molecular Formula: C28H32ClN3O8S
InChi Key: DSKIOWHQLUWFLG-SPIKMXEPSA-N
SMILES: OC(=O)\C=C/C(O)=O.OC(=O)\C=C/C(O)=O.CN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(Cl)C=C23)CC1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results