accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 81636-17-1

CH3NHOBr

CAS RN 81636-17-1

IUPAC Name: 4-bromo-N-(3-methylphenyl)benzamide
Synonyms: 4-bromo-N-(3-methylphenyl)benzamide
Molecular Weight (g/mol): 290.16
Molecular Formula: C14H12BrNO
InChi Key: MRYWJNCVXBZMKZ-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=CC=C(Br)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results