accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 81025-04-9

OOHOHOOHHOOHOHHOHOOHH2O

CAS RN 81025-04-9

IUPAC Name: 4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol hydrate
Synonyms: maltitol hydrate
Molecular Weight (g/mol): 362.33
Molecular Formula: C12H26O12
InChi Key: LXMBXZRLTPSWCR-UHFFFAOYNA-N
SMILES: O.OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)