accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79803-73-9

ONSOOOHONa

CAS RN 79803-73-9

IUPAC Name: sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
Synonyms: sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
Molecular Weight (g/mol): 247.24
Molecular Formula: C7H14NNaO5S
InChi Key: WSFQLUVWDKCYSW-UHFFFAOYNA-M
SMILES: [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (1)
pH 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)