accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79418-78-3

CH3OOFOH

CAS RN 79418-78-3

IUPAC Name: 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
Synonyms: 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: OOGOFUKAJDPHDJ-UHFFFAOYSA-N
SMILES: COC1=C(O)C(F)=CC(C=O)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results