accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 78-44-4

H3CCH3ONH2OOOHNH3CCH3

CAS RN 78-44-4

IUPAC Name: 2-[(carbamoyloxy)methyl]-2-methylpentyl N-(propan-2-yl)carbamate
Synonyms: carisoprodol Soma Rela chinchen vanadom carisoma carisol somadril flexal soprodol
Molecular Weight (g/mol): 260.33
Molecular Formula: C12H24N2O4
InChi Key: OFZCIYFFPZCNJE-UHFFFAOYNA-N
SMILES: CCCC(C)(COC(N)=O)COC(=O)NC(C)C
Industry Type 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results