CAS RN 779353-01-4
CAS RN 779353-01-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Dinaciclib
Potent and selective inhibitor of CDK2, CDK5, CDK1 and CDK9
2
Dinaciclib, MedChemExpress
MedChemExpress Dinaciclib (SCH 727965) is a potent inhibitor of CDK, with IC50s of 1 nM, 1 nM, 3 nM, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively.