Learn More
Dinaciclib, MedChemExpress
Product Code. 16450357
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16450357
Brand: Medchem Express HY10492
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Dinaciclib (SCH 727965) is a potent inhibitor of CDK, with IC50s of 1 nM, 1 nM, 3 nM, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively.
Dinaciclib (SCH 727965) is a potent inhibitor of CDK, with IC50s of 1 nM, 1 nM, 3 nM, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively.
Specifications
Chemical Name or Material Dinaciclib
Physical Form Solid
CAS 779353-01-4
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H28N6O2
For Use With (Application) Cancer-Kinase/protease
Synonym SCH 727965
Solubility Information DMSO : 50 mg/mL (126.11 mM; Need ultrasonic)
SMILES OCC[C@H]1N(CCCC1)C2=NC3=C(C=NN3C(NCC4=C[N+]([O-])=CC=C4)=C2)CC
Molecular Weight (g/mol) 396.49
Formula Weight 396.49
Percent Purity 95.75%
Grade Research