accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 75607-67-9

N(R)N(S)O(S)(R)NNOHNH2HOOFPOOHHO

CAS RN 75607-67-9

IUPAC Name: {[5-(6-amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: fludarabine
Molecular Weight (g/mol): 365.21
Molecular Formula: C10H13FN5O7P
InChi Key: GIUYCYHIANZCFB-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(COP(O)(O)=O)C(O)C3O)C2=NC(F)=N1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results