accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

LiH3CCH3ClMgCl

CAS RN 745038-86-2

IUPAC Name: magnesium(2+) lithium(1+) propan-2-ide dichloride
Synonyms: magnesium(2+) lithium(1+) propan-2-ide dichloride
Molecular Weight (g/mol): 145.23
Molecular Formula: C3H7Cl2LiMg
InChi Key: DBTNVRCCIDISMV-UHFFFAOYSA-L
SMILES: [Li+].[Mg++].[Cl-].[Cl-].C[CH-]C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results