accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 74257-00-4

IUPAC Name: 3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Synonyms: 3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1,2,4-triazole
Molecular Weight (g/mol): 296.3
Molecular Formula: C11H12N4O4S
InChi Key: SFYDWLYPIXHPML-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results