accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3NOHHO

CAS RN 73708-65-3

IUPAC Name: 1,3-dihydroxypropan-2-aminium
Synonyms: 1,3-dihydroxypropan-2-aminium
Molecular Weight (g/mol): 92.12
Molecular Formula: C3H10NO2
InChi Key: KJJPLEZQSCZCKE-UHFFFAOYSA-O
SMILES: [NH3+]C(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results