accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 73259-81-1

H3CCH3CH3OONH(S)NH3OO

CAS RN 73259-81-1

IUPAC Name: (2S)-3-azaniumyl-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2S)-3-ammonio-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 204.23
Molecular Formula: C8H16N2O4
InChi Key: KRJLRVZLNABMAT-YFKPBYRVSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](C[NH3+])C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)