accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7311-68-4

SBrOHOBr

CAS RN 7311-68-4

IUPAC Name: 3,5-dibromothiophene-2-carboxylic acid
Synonyms: 3,5-dibromothiophene-2-carboxylic acid
Molecular Weight (g/mol): 285.94
Molecular Formula: C5H2Br2O2S
InChi Key: YWOSIPRAZKDEIL-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results