accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7305-71-7

CH3SNH2N

CAS RN 7305-71-7

IUPAC Name: 5-methyl-1,3-thiazol-2-amine
Synonyms: 5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: GUABFMPMKJGSBQ-UHFFFAOYSA-N
SMILES: CC1=CN=C(N)S1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results