accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 725743-50-0

CH3FFFOCH3

CAS RN 725743-50-0

IUPAC Name: 1-[2-methyl-5-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[2-methyl-5-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 202.18
Molecular Formula: C10H9F3O
InChi Key: ZLDDSCHHMXTRHJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results