CAS RN 72235-58-6
CAS RN 72235-58-6
IUPAC Name:
1-(4-chloro-3-fluorophenyl)methanamine
Synonyms:
1-(4-chloro-3-fluorophenyl)methanamine
Molecular Weight (g/mol):
159.59
Molecular Formula:
C7H7ClFN
InChi Key:
HSNPBYKCCNMQNA-UHFFFAOYSA-N
SMILES:
NCC1=CC=C(Cl)C(F)=C1
1
–
1
of
1
results
1
4-Chloro-3-fluorobenzylamine, 97%, Thermo Scientific™
CAS: 72235-58-6 Molecular Formula: C7H7ClFN Molecular Weight (g/mol): 159.588 MDL Number: MFCD04116008 InChI Key: HSNPBYKCCNMQNA-UHFFFAOYSA-N Synonym: 4-chloro-3-fluorophenyl methanamine,4-chloro-3-fluorobenzylamine,4-chloro-3-fluorobenzyl amine,benzenemethanamine,4-chloro-3-fluoro,benzenemethanamine, 4-chloro-3-fluoro,4-chloro-3-fluorophenyl methylamine,1-4-chloro-3-fluorophenyl methanamine,3-fluoro-4-chlorobenzyl amine,rarechem al bw 1597,4-chloranyl-3-fluoranyl-phenyl methanamine PubChem CID: 2757541 IUPAC Name: (4-chloro-3-fluorophenyl)methanamine SMILES: C1=CC(=C(C=C1CN)F)Cl