accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 72235-51-9

IUPAC Name: (2,3-difluorophenyl)methanaminium
Synonyms: (2,3-difluorophenyl)methanaminium
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H8F2N
InChi Key: OHZUCDHZOHSBPZ-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=CC(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results