CAS RN 718635-93-9
CAS RN 718635-93-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetrabenazine Racemate, MedChemExpress
MedChemExpress Tetrabenazine Racemate (Ro 1-9569 Racemate) is a selective and reversible inhibitor of vesicular monoamine transporter-2 (VMAT-2).