accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7081-44-9

H2ONaH3CONClOHN(R)(R)SCH3CH3(S)NOOO

CAS RN 7081-44-9

IUPAC Name: sodium (2S,5R,6R)-6-[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate
Synonyms: tegopen cloxapen cloxacillin sodium hydrate sodium cloxacillin monohydrate cloxacillin sodium monohydrate bactopen
Molecular Weight (g/mol): 475.88
Molecular Formula: C19H19ClN3NaO6S
InChi Key: KCUWTKOTPIUBRI-VICXVTCVSA-M
SMILES: O.[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)