CAS RN 70459-04-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Formyl Norfloxacin, TRC
CAS: 70459-04-0 Molecular Formula: C17 H18 F N3 O4 Molecular Weight (g/mol): 347.34 Synonym: 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid,Norfloxacin Imp. G (EP) IUPAC Name: 1-ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid SMILES: CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(CC3)C=O
2
1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.