accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 689254-23-7

H3COBrFF

CAS RN 689254-23-7

IUPAC Name: 1-(bromomethyl)-2,3-difluoro-4-methoxybenzene
Synonyms: 1-(bromomethyl)-2,3-difluoro-4-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: AMLNZEUOTXDYNB-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results