accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

KH3CH3CO

CAS RN 6831-82-9

IUPAC Name: potassium propan-2-olate
Synonyms: potassium propan-2-olate
Molecular Weight (g/mol): 98.19
Molecular Formula: C3H7KO
InChi Key: WQKGAJDYBZOFSR-UHFFFAOYSA-N
SMILES: [K+].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results