accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68099-86-5

HClH3CCH3ONN

CAS RN 68099-86-5

IUPAC Name: hydrogen N-benzyl-N-[3-(2-methylpropoxy)-2-(pyrrolidin-1-yl)propyl]aniline chloride
Synonyms: bepridil hydrochloride
Molecular Weight (g/mol): 403.01
Molecular Formula: C24H35ClN2O
InChi Key: JXBBWYGMTNAYNM-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)COCC(CN(CC1=CC=CC=C1)C1=CC=CC=C1)N1CCCC1
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results