Learn More
Bepridil hydrochloride, MedChemExpress
Product Code. 16496547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16496547
Brand: Medchem Express HY103315
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity.
Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity.
Specifications
Chemical Name or Material Bepridil hydrochloride
Physical Form Solid
CAS 68099-86-5
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H35ClN2O
For Use With (Application) COVID-19-immunoregulation
Synonym CERM 1978
Solubility Information DMSO : ≥ 150 mg/mL (372.21 mM) ∣H2O : 12.5 mg/mL (31.02 mM; Need ultrasonic)
SMILES [H]Cl.CC(C)COCC(N1CCCC1)CN(C2=CC=CC=C2)CC3=CC=CC=C3
Molecular Weight (g/mol) 403.0
Formula Weight 403.0
Percent Purity 99.25%
Grade Research