accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CMgCl

CAS RN 676-58-4

IUPAC Name: methylmagnesium chloride
Synonyms: methylmagnesium chloride
Molecular Weight (g/mol): 74.79
Molecular Formula: CH3ClMg
InChi Key: CCERQOYLJJULMD-UHFFFAOYSA-M
SMILES: [CH3-].[Mg++].[Cl-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results