accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 676-46-0

IUPAC Name: disodium 2-hydroxybutanedioate
Synonyms: sodium malate
Molecular Weight (g/mol): 178.05
Molecular Formula: C4H4Na2O5
InChi Key: WPUMTJGUQUYPIV-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results