accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67533-12-4

CH3NH3CH3CH3CCH3OOH2OH2OH2OH2O

CAS RN 67533-12-4

IUPAC Name: tetraethylazanium acetate tetrahydrate
Synonyms: tetraethylammonium acetate tetrahydrate
Molecular Weight (g/mol): 261.36
Molecular Formula: C10H31NO6
InChi Key: QNOAKQGNSWGYOE-UHFFFAOYSA-M
SMILES: O.O.O.O.CC([O-])=O.CC[N+](CC)(CC)CC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)