accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67515-55-3

OOFFFF

CAS RN 67515-55-3

IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzoate
Synonyms: 4-fluoro-3-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 207.1
Molecular Formula: C8H3F4O2
InChi Key: WZBPZYCJUADXRS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results