accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SNHO

CAS RN 6670-13-9

IUPAC Name: 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione
Synonyms: 4,5-diphenyl-3H-1,3-oxazole-2-thione
Molecular Weight (g/mol): 253.32
Molecular Formula: C15H11NOS
InChi Key: NYQSTRUSYDZNHC-UHFFFAOYSA-N
SMILES: S=C1NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results