accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6590-91-6

IUPAC Name: (2,6-dichlorophenyl)thiourea
Synonyms: 2,6-dichlorophenylthiourea
Molecular Weight (g/mol): 221.1
Molecular Formula: C7H6Cl2N2S
InChi Key: KUQHRGMPBWZVQR-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results