accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HClClNHN

CAS RN 65369-76-8

IUPAC Name: hydrogen 1-(3-chlorophenyl)piperazine chloride
Synonyms: hydrogen m-chlorophenylpiperazine chloride
Molecular Weight (g/mol): 233.14
Molecular Formula: C10H14Cl2N2
InChi Key: MHXPYWFZULXYHT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=CC(=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results