accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOFFFFOO

CAS RN 652-03-9

IUPAC Name: tetrafluorobenzene-1,2-dicarboxylate
Synonyms: tetrafluorophthalate
Molecular Weight (g/mol): 236.08
Molecular Formula: C8F4O4
InChi Key: YJLVXRPNNDKMMO-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=C(F)C(F)=C(F)C(F)=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results