CAS RN 64464-07-9
CAS RN 64464-07-9
IUPAC Name:
2-(2-methoxyphenoxy)ethan-1-amine hydrochloride
Synonyms:
2-(2-methoxyphenoxy)ethanamine hydrochloride
Molecular Weight (g/mol):
203.67
Molecular Formula:
C9H14ClNO2
InChi Key:
KNWPXZOMSZABHD-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=CC=C1OCCN
1
–
1
of
1
results
1
2-(2-Methoxyphenoxy)ethylamine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.