accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6411-75-2

H3CCH3NH3H3CCH3NH3OSOOO

CAS RN 6411-75-2

IUPAC Name: bis(heptan-2-aminium) sulfate
Synonyms: tuaminoheptane sulfate
Molecular Weight (g/mol): 328.51
Molecular Formula: C14H36N2O4S
InChi Key: XKUUMWKWUZRRPD-UHFFFAOYNA-N
SMILES: [O-]S([O-])(=O)=O.CCCCCC(C)[NH3+].CCCCCC(C)[NH3+]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results