accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6335-39-3

IUPAC Name: 4-(propan-2-yl)benzene-1-sulfonamide
Synonyms: 4-isopropylbenzenesulfonamide
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: WVOWEROKBOQYLJ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results