accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62929-02-6

A(a)

CAS RN 62929-02-6

IUPAC Name: 1-(4-aminophenyl)-1,3,3-trimethyl-2,3-dihydro-1H-inden-5-amine; 5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl)-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine; 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 588.62
Molecular Formula: C35H28N2O7
InChi Key: CDTDIQLZIBORMV-UHFFFAOYNA-N
SMILES: CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)