accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6285-05-8

CH3OCl

CAS RN 6285-05-8

IUPAC Name: 1-(4-chlorophenyl)propan-1-one
Synonyms: 1-(4-chlorophenyl)propan-1-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: ADCYRBXQAJXJTD-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)