accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62649-23-4

NH2On

CAS RN 62649-23-4

IUPAC Name: sodium prop-2-enamide prop-2-enoic acid prop-2-enoate
Synonyms: sodium acrylic acid polyacrylamide acrylate
Molecular Weight (g/mol): 237.19
Molecular Formula: C9H12NNaO5
InChi Key: DTBMFUOWAASNDB-UHFFFAOYSA-M
SMILES: [Na+].NC(=O)C=C.OC(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results