accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 626-27-7

H3COOOCH3

CAS RN 626-27-7

IUPAC Name: heptanoyl heptanoate
Synonyms: heptanoic acid, anhydride
Molecular Weight (g/mol): 242.36
Molecular Formula: C14H26O3
InChi Key: DAPZDAPTZFJZTO-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)OC(=O)CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)