accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 625-98-9

IUPAC Name: 1-chloro-3-fluorobenzene
Synonyms: 1-chloro-3-fluorobenzene
Molecular Weight (g/mol): 130.55
Molecular Formula: C6H4ClF
InChi Key: VZHJIJZEOCBKRA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results