accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOOHN

CAS RN 62497-65-8

IUPAC Name: 4-nitro-N-(2-phenylethyl)benzamide
Synonyms: 4-nitro-N-(2-phenylethyl)benzamide
Molecular Weight (g/mol): 270.29
Molecular Formula: C15H14N2O3
InChi Key: NPTZNBFQWMQHKP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)NCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results