CAS RN 623-95-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Chlorpropamide EP Impurity B, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N,N’-Dipropylurea, TRC
CAS: 623-95-0 Molecular Formula: C7 H16 N2 O Molecular Weight (g/mol): 144.215 Synonym: N,N'-Dipropylurea,Urea, 1,3-dipropyl- (6CI,7CI,8CI),1,3-Di-n-propylurea,1,3-Dipropylurea,N,N'-Dipropylurea,Chlorpropamide Imp. B (EP 7.0) IUPAC Name: 1,3-dipropylurea SMILES: CCCNC(=O)NCCC