accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 621-56-7

H3CNCH3HOOH

CAS RN 621-56-7

IUPAC Name: 3-(diethylamino)propane-1,2-diol
Synonyms: 3-(diethylamino)propane-1,2-diol
Molecular Weight (g/mol): 147.22
Molecular Formula: C7H17NO2
InChi Key: LTACQVCHVAUOKN-UHFFFAOYNA-N
SMILES: CCN(CC)CC(O)CO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)