accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62059-43-2

OOOHOOHH2O

CAS RN 62059-43-2

IUPAC Name: 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid hydrate
Synonyms: endothal hydrate
Molecular Weight (g/mol): 204.18
Molecular Formula: C8H12O6
InChi Key: RHLALKDQINFLPM-UHFFFAOYNA-N
SMILES: O.OC(=O)C1C2CCC(O2)C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results